Learn More
3M™ Inhalation Valve
3M™ Inhalation Valve is suitable for use with 3M™ Reusable Half Mask 7500 Series.
Brand: 3M™ 7000029731
46.03 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Replacement inhalation valve for half mask respirator
- Replace damaged parts to ensure correct levels of protection and comfort
- For use with 3M™ Half Face Masks 7500 Series
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Light Blue | |
Inhalation Valve | |
For all painting work, mixing of paints and use of adhesives | |
Half Mask Series 7500 |