missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Injection Needle
With O.D. 0.7mm guidance
Brand: Cytiva 18180142
This item is not returnable.
View return policy
Description
- With guidance of 0.7mm O.D.

Specifications
Injection Needle | |
0.7 mm O.D. guidance |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytiva Injection Needle >
Spot an opportunity for improvement?Share a Content Correction