missing translation for 'onlineSavingsMsg'
Learn More

Miele Professional™ Injector nozzle

Injector nozzles ID 200 with plastic supports, Ø 6, length 200 mm

Brand:  Miele Professional™ 3810380

Product Code. 10201622

  • 34.50 € / Each
Estimated Shipment: 11-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Suitable for standard mobile injector units and injector baskets
  • Suitable for measuring flasks and test tubes, for example
Specifications

Specifications

Injector nozzle
Stainless Steel, Black
200 mm
Stainless Steel, Plastic
Washer-disinfectors, laboratory
6 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.