missing translation for 'onlineSavingsMsg'
Learn More

B Braun™ Injekt™ Solo Disposable Centric Luer Two-Part Syringe

Manufactured from polypropylene/polyethylene. B. Braun™ Injekt™ Solo Disposable Centric Luer-lock Two-Part Syringe has high contrast graduations for ideal readability.

36.00 €

Specifications

Certifications/Compliance ISO 7886-1
Material Polyethylene, Polypropylene
Color Green Plunger
Description Black graduation
For Use With (Application) Ideal for simple injections, vaccinations and more
View More Specs

Products 1
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
12762637
View Documents
B Braun™
460670-1V
2 mL
36.00 €
Pack of 100
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Made of polypropylene/polyethylene
  • „Green plunger
  • „Highly transparent barrel
  • „High contrast black graduation, permanent marking, ideal readability
  • „Safe plunger backstop
  • „Easy aspiration up to maximum volume
  • „Latex-free/PVC-free
  • „Silicone oil-free
  • Single sterile-packed (in box of 100 pcs)
Specifications

Specifications

ISO 7886-1
Green Plunger
Ideal for simple injections, vaccinations and more
Centric Luer-lock 2-part Syringe
100/Pk.
Yes
Polyethylene, Polypropylene
Black graduation
Yes
Single-use
Centric Luer Lock
Sterile
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 5 Years

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
B Braun™ Injekt™ Solo Disposable Centric Luer Two-Part Syringe

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.