missing translation for 'onlineSavingsMsg'
Learn More
Learn More
METTLER TOLEDO™ InLab™ Easy pH Electrodes
A durable, polysulfone pH electrode with an integral bulb protector and gel electrolyte to avoid the inconvenience of refilling in field. Temperature sensors sold separately.
Brand: METTLER TOLEDO™ 51343010
127.20 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.
This item is not returnable.
View return policy
Description
- For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
- Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
- Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.
Featured Applications
- Aquarium water
- Soft drinks
- Beer
- General lab
Accessories & Care
- Ceramic junction
- Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
- Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
- Temperature sensors sold separately (NTC 30 kΩ: 51300164, PT1000: 51300165)
Specifications
S7 | |
Polysulfone | |
0°C to 80°C | |
Gel | |
S7 to BNC (30281915) | |
U Glass | |
NTC 30 kilohm | |
3mol/L KCl | |
0°C to 80°C | |
120 mm | |
0.47 in. |
Ceramic | |
0 to 14 | |
Combination Electrode | |
Low maintenance, field measurement and educational use | |
Ag/AgCl | |
InLab™ Easy | |
<20 sec. | |
32°F to 176°F | |
4.72 in. | |
w/out Cable | |
12 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction