missing translation for 'onlineSavingsMsg'
Learn More

METTLER TOLEDO™ InLab™ Expert pH Electrodes

Product Code. 10447144 Shop All Mettler Toledo Products
Click to view available options
Connector Types:
BNC, RCA
BNC, RCA IP67
DIN 19262, 4 mm Banana Plug
Multi-Pin
S7
Includes:
Electrode only
ISM Temperature Probe
Pt1000 Temperature Probe
Temperature Probe
Temperature Probe (NTC30)
Length (Metric) Cable:
1.2 m
1.8 m
10 m
2 m
No Cable
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10447144

Brand: METTLER TOLEDO™ 51340288

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

A combination pH electrode with PEEK plastic shaft for chemical and mechanical durability, and a fixed cable. Designed for use in a variety of applications.

Ideal for critical samples such as suspensions or samples of unknown composition. Mettler Toledo Expert Series pH and pH/ATC Combination Electrodes with open junction puts the reference electrode in direct contact with the measured medium, eliminating the possibility of contamination or blockage.

  • For use with METTLER TOLEDO SevenExcellence, SevenCompact and portable meters as well as other brands. Connection cables may be required.
  • Features a PEEK plastic shaft for harsh environments and requires no maintenance
  • Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
  • Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.

Featured Applications

  • General Lab
  • Beverages
  • Waste water
  • Suspended solids
  • Environmental
  • TRIS compatible

Accessories & Care

  • Open Junction and non-refillable - no additional electrolyte required
  • Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
  • Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
TRUSTED_SUSTAINABILITY

Specifications

Connector Types BNC, RCA IP67
Junction Ceramic
Material PEEK
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 100°C
Type Combination Electrode, Integrated Temperature Probe
Filling Solution Type Xerolyt Polymer
For Use With (Application) Emulsions, suspensions, effluent water and solutions containing Proteins and Sulphides
For Use With (Equipment) Fixed Cable to sensor
Includes Temperature Probe
Internal Reference ARGENTHAL
Membrane U Glass
Model InLab Expert Go
Probe NTC 30 kilohm
Response Time <20 sec.
Storage Requirements FRISCOLYT-B
Temperature (English) Operating 32°F to 212°F
Temperature (Metric) Range 0°C to 60°C
Length (English) 4.7 in.
Length (English) Cable 78.7 in.
Length (Metric) 120 mm
Length (Metric) Cable 2 m
Diameter (English) 0.47 in.
Diameter (Metric) 12 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.