missing translation for 'onlineSavingsMsg'
Learn More
Learn More
METTLER TOLEDO™ InLab™ Routine pH Electrodes
A combination pH electrode with an integrated temperature sensor and chemically resistant HA glass body. It is designed for general lab applications and ideal for use at high pH values (IP67 rated).
236.00 € - 503.00 €
Specifications
Junction | Ceramic |
---|---|
Material | Glass |
pH Range | 0 to 14 |
Temperature (Metric) Operating | 0°C to 100°C |
Membrane | HA Glass |
Product Code | Brand | Model | Type | Connector Types | For Use With (Equipment) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Type | Connector Types | For Use With (Equipment) | Price | Quantity & Availability | ||
10650324
|
METTLER TOLEDO™
51343054 |
InLab™ Routine Pro | Combination Electrode, Integrated Temperature Probe | Multi-Pin | Multi-Pin to BNC (30281896) |
343.00 €
Each |
In Stock
Log in to see stock. |
||
10313433
|
METTLER TOLEDO™
51343050 |
InLab™ Routine | Combination Electrode | S7 | S7 to BNC (30281915) |
236.00 €
Each |
In Stock
Log in to see stock. |
||
10212283
|
METTLER TOLEDO™
51343056 |
InLab™ Routine Pt1000 | Combination Electrode; Integrated Temperature Probe (Pt1000) | Multi-Pin | Multi-Pin to BNC (30281896) |
343.00 €
Each |
In Stock
Log in to see stock. |
||
11354213
|
METTLER TOLEDO™
51344055 |
InLab™ Routine Pro-ISM | Combination Electrode, Integrated Temperature Probe, Intelligent Sensor Management (ISM) Technology | Multi-Pin | SevenCompact pH/ion meter |
421.00 €
Each |
Estimated Shipment: 14-02-2025
Log in to see stock. |
||
15529611
|
METTLER TOLEDO™
30248832 |
InLab™ Routine Go-ISM | Combination Electrode, Integrated Temperature Probe, Intelligent Sensor Management (ISM) Technology | BNC IP67 | Fixed Cable to sensor |
503.00 €
Each |
In Stock
Log in to see stock. |
||
Description
- For use with METTLER TOLEDO SevenExcellence, SevenCompact and portable meters as well as other brands. Connection cables may be required.
- Intelligent Sensor Management (ISM) electrodes capture and save calibration data, which is then automatically recognized by the meter once connected.
- Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
- Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.
Featured Applications
- Drinking water
- Highly saline solutions or seawater
- Microbiological samples
- Corrosive acids and bases
- General Lab
- Chemical
- High pH values
- TRIS compatible
Accessories & Care
- Refillable 3M KCl (51350072): Keep probe filled with electrolyte for accurate results
- Tip: Open fill port while measuring, and also to refill electrolyte solution
- Ceramic junction
- Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
- Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
Specifications
Ceramic | |
0 to 14 | |
HA Glass |
Glass | |
0°C to 100°C | |
0°C to 100°C |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title