missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ InLab™ Sensor Cables: MultiPin™ Connection

Electrode cables to connect Mettler Toledo InLab sensors to any Mettler Toledo meter with compatible connector.

Brand:  Mettler Toledo™ 30281899

 View more versions of this product

Product Code. 15519651

  • 132.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Detachable
  • Resuable
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MultiPin-BNC/1 x 4mm banana
To connect Mettler Toledo InLab sensors to any meter
1.2 m
Mettler Toledo InLab Sensors
4 ft.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.