missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mettler Toledo™ InLab™ Sensor Cables: MultiPin™ Connection
Electrode cables to connect Mettler Toledo InLab sensors to any Mettler Toledo meter with compatible connector.
Brand: Mettler Toledo™ 30281913
138.12 EUR valid until 2025-03-30
Use promo code "24134" to get your promotional price.
This item is not returnable.
View return policy
Description
- Detachable
- Resuable
Specifications
MultiPin-BNC/RCA (cinch), IP67 waterproof | |
To connect Mettler Toledo InLab sensors to any meter | |
1.8 m |
Mettler Toledo InLab Sensors | |
6 ft. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction