Learn More
Welch™ Inlet Separator for CRVpro Direct Drive Rotary Vane Vacuum Pump
For use with CRVpro Direct Drive Rotary Vane Vacuum Pump
Brand:  Welch™ 320016
Product Code. 10753922
  • 286.00 € / Each
Estimated Shipment: 24-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Intake separator separates liquid phase aerosols from gas stream, reducing chemical contaminants in pump oil
Specifications
Specifications
Inlet Separator
Separates liquid phase aerosols from gas stream
Rotary Vane Pump