missing translation for 'onlineSavingsMsg'
Learn More

Cole Parmer™ Inoculating Loop, Platinum

Precisely manufactured platinum and iridium wire for transferring bacterial cultures.

Brand:  Cole Parmer™ 01850-04

Product Code. 11719679

  • 223.00 € / Each
Estimated Shipment: 17-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Platinum loop ideal for transferring bacterial cultures.
  • Constructed of Bamp;S platinum wire with 15% iridium for rigidity - this allows for a thinner wire for delicate work.
  • Loops are fused to form a perfect circle with an inside tolerance of ±0.02 mm to ensure accurate delivery.
  • Order insulated loop and holder separately.
Specifications

Specifications

2 mm
45 mm
Platinum Iridium
24
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers