Learn More
Kartell™ Inoculation Loops and Needles
Manufactured with high impact polystyrene for good dimensional stability. Kartell™ Inoculation Loops and Needles provide a two–in–one sterile disposable tool for inoculation.
Brand:  Kartell™ 0265000
Product Code. 10789651
  • ONLINE EXCLUSIVE PRICE
    605.00 €
    450.75 € / Pack of 8000
    Save 154.25 €
    25% Off
Only null left
Estimated Shipment: 16-03-2026
Add to basket
Description
Description
  • Gamma sterilized and packed in polybags
  • Looped end to retrieve an inoculum from a culture, transfer inoculum for streaking or transfer microscopic organisms
  • Needle end for picking off individual isolated colonies  or making stab inoculations into agar slants or tubes of solid culture medium
  • CE–IVD