missing translation for 'onlineSavingsMsg'
Learn More

WTW™ inoLab™ Cond 7310 Precise and Convenient Conductivity Meter
GREENER_CHOICE

A precise and high performance laboratory conductivity meter with GLP (Good Laboratory Practice) and AQA (Analytical Quality Assurance) supporting functions for documentation measuring results.

Brand:  WTW™ 1CA301

1647.00 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.


Product Code. 12691685

  • 2065.00 € / Each
Estimated Shipment: 16-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Documentation of all data via USB interface or optional built-in printer: fast and error-free
  • Suitable for measurements according to pharmacopeia
  • For documentation, the serial number of the used sensor can be entered
  • Repeatable measuring results by active, automatic AutoRead function with automatic recognition of stable values
  • The sensor symbol informs about the status of the electrode
  • Graphic display with plain text menus for convenient and safe operation
  • Includes the MultiLab™ Importer to import data via Excel™
  • For AC and battery operation
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Meter with universal power supply, stand and operation manual, including 4-electrode graphite conductivity cell TetraCon™ 325, 0.01mol/L KCl conductivity standard, software and USB cable
9V, 1.1A
1
nLF, linear 0.000 to 10.000%, can be switched-off
0 to 1999 mg/L
cETLus, CE
inoLab™ Cond 7310
-5°C to +105°C
100 to 240V, 50/60Hz, 270mA input, 9V output
50/60Hz
±0.5% of value
Fixed 0.01cm–1, calibration 0.450 to 0.500cm–1, 0.800 to 0.880cm–1, adjustable 0.09 to 0.110cm–1, 0.250 to 25cm–1
0 μS/cm to 1000 mS/cm
0 to 70 ppt
100 to 240V, 50 to 60Hz, 270mA
0.8 kg
4 x 1.5V Alkali Manganese batteries, Type AA, 4 x 1.2V NiMH rechargeable batteries, type AA
Precise and Convenient Conductivity Meter
100/240V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.