Learn More
iNOS Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22226-1-AP-20UL
This item is not returnable.
View return policy
Description
The iNOS antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human iNOS. This antibody recognizes human, mouse antigen. The iNOS antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
iNOS | |
Polyclonal | |
Unconjugated | |
20 μL | |
4843 | |
IgG |
Western Blot, Immunohistochemistry, Immunofluorescence | |
533 μg/mL | |
Rabbit | |
Primary | |
Human, Mouse |