Learn More
Thermo Scientific Chemicals Inositol hexaphosphoric acid, 40-50 wt.% aqueous solution
Brand: Thermo Scientific Chemicals 235370025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Inositol hexaphosphoric acid | |
83-86-3, 7732-18-5 | |
C6H18O24P6 | |
1.3700g/mL | |
Solubility in water: miscible. | |
OP(O)(=O)O[C@H]1[C@H](OP(O)(O)=O)[C@@H](OP(O)(O)=O)[C@H](OP(O)(O)=O)[C@H](OP(O)(O)=O)[C@@H]1OP(O)(O)=O | |
660.03 | |
660.03 | |
1.391 | |
2.5 L |
40-50 wt. % Aqueous Solution | |
105.0°C | |
MFCD00082309 | |
phytic acid, phytate, fytic acid, inositol hexaphosphate, alkalovert, myo-inositol hexaphosphate, alkovert, phytine, acide fytique, acidum fyticum | |
IMQLKJBTEOYOSI-GPIVLXJGSA-N | |
(2,3,4,5,6-pentaphosphonooxycyclohexyl) dihydrogen phosphate | |
890 | |
Glass bottle | |
1.37 |
Safety and Handling
- Inositol hexaphosphoric acid, 40-50 wt% aqueous solution
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- H290-May be corrosive to metals.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only