missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Miele Professional™ Insert
454.00 € - 530.00 €
Product Code | Brand | Height (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Height (Metric) | Price | Quantity & Availability | ||||||
13112280
|
Miele Professional™
3830290 |
95 mm |
530.00 €
Each |
Estimated Shipment: 20-06-2025
Log in to see stock. |
||||||
13192270
|
Miele Professional™
3830270 |
120 mm |
454.00 €
Each |
Estimated Shipment: 14-04-2025
Log in to see stock. |
||||||
13102280
|
Miele Professional™
3830280 |
145 mm |
530.00 €
Each |
Estimated Shipment: 20-06-2025
Log in to see stock. |
||||||
Description
Specifications
Insert | |
445 mm | |
1 Each |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Miele Professional™ Insert