Learn More
Miele Professional™ Insert
Mobile unit E 960/1 for the optimum loading of wide necked Erlenmeyer flasks
Brand:  Miele Professional™ 5892360
Product Code. 15430565
  • 1485.00 € / Each
Estimated Shipment: 02-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Equipped with 20 large and 26 small spring hooks
  • Suitable for Erlenmeyer flasks and measuring cylinders
Specifications
Specifications
Stainless Steel
Stainless Steel
Large-capacity washer-disinfectors, laboratory
185 mm
Insert/Module for Wide-necked Glassware
46 Erlenmeyer flasks
522 mm
357 mm