missing translation for 'onlineSavingsMsg'
Learn More

Miele Professional™ Insert

Insert E 969 for holding different utensils

Brand:  Miele Professional™ 5746240

Product Code. 15470565

  • 201.00 € / Each
Estimated Shipment: 11-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Perforated plate 7 x 7 x 3 mm
  • Suitable for E 900-4/1, E 935/1, E 975/2, E 941 with E 945
Specifications

Specifications

Stainless Steel
Stainless Steel
533 mm
363 mm
Insert/Module for Beakers
Large-capacity washer-disinfectors, laboratory
122 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.