Learn More
Miele Professional™ Insert
E 750 frames for the optimum securing of different inserts
Brand:  Miele Professional™ 5567510
Product Code. 15470545
  • 430.00 € / Each
Estimated Shipment: 26-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Holding frames for securing inserts E 106 to E 111
  • Enables E 135 insert for baby bottles to be loaded in an E 555/2
  • Enables E 135 insert for baby bottles to be loaded in an E 735/2
Specifications
Specifications
Stainless Steel
Stainless Steel
Washer-disinfectors, laboratory