missing translation for 'onlineSavingsMsg'
Learn More

Miele Professional™ Insert

Product Code. 10170641 Shop All Miele Products
Click to view available options
:
Dimensions (D x W):429 x 224mm
This item is not returnable. View return policy

Product Code. 10170641

Brand: Miele Professional™ 6629670

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Insert A12/1 for the gentle cleaning of delicate items by interrupting the spray jet

  • With perforations 7 x 7 mm
  • For use in upper and lower basket
TRUSTED_SUSTAINABILITY

Specifications

Material Stainless Steel
Color White
For Use With (Equipment) Washer-disinfectors, laboratory
Depth (Metric) 429 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.