Learn More
Miele Professional™ Insert
Mobile unit E 340 for the optimum loading of narrow necked glassware
Brand:  Miele Professional™ 3809410
1030.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10560672
  • 1110.00 € / Each
Estimated Shipment: 27-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Half can be equipped with other inserts
  • Equipped with 19 injector nozzles
Specifications
Specifications
Stainless Steel
Stainless Steel
Washer-disinfectors, laboratory
502 mm
Mobile Unit for Narrow-necked Glassware
19 Narrow-necked glassware
517 mm
535 mm