missing translation for 'onlineSavingsMsg'
Learn More

Miele Professional™ Insert

Insert E 106 for the optimum loading of wide neck flasks, etc

Brand:  Miele Professional™ 3808310

219.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 10019350

  • 232.00 € / Each
Estimated Shipment: 27-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Equipped with 10 spring hooks x 175 mm height & 16 x 105mm height
  • The distance between the spring hooks is approx. 60 mm
Specifications

Specifications

Insert, Half
Wide-necked Glassware, Measuring Beakers, etc
430 mm
195 mm
E 106
10 large spring hooks x 175mm height (distance between spring hooks is approx. 60mm)
186 mm
1 Each
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.