missing translation for 'onlineSavingsMsg'
Learn More

Miele Professional™ Insert

Insert E 110 for the optimum loading of beakers from 250 to 600 ml

Brand:  Miele Professional™ 3808390

Product Code. 10384531

  • 237.00 € / Each
Estimated Shipment: 07-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Suitable for 10 glass beakers between 250 - 600 ml
  • equipped with 10 x 3 stay bars
  • For use lower basket
Specifications

Specifications

Insert, Half
10 Beakers, 250 to 600mL
175 mm
1 Each
E 110
460 mm
230 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.