Learn More
Fisherbrand™ Insert For Maxi Boxes
Manufactured with hinged lids and accommodate interchangeable inserts that hold microtubes, centrifuge tubes, scintillation vials, universals, cryotubes and falcon tubes; these serve as an ideal solution for short-term storage of radioactive waste or radioisotopes.
Brand:  Fisherbrand™ CSR-R15
 View more versions of this product
Product Code. 10315311
  • 46.80 € / Each
Estimated Shipment: 11-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Designed to hold up to three midi-box inserts, as well as additional inserts for taller types of tubes. Available for either beta or gamma radiation protection
  • Cleaver Scientific storage boxes are manufactured with hinged lids and accommodate interchangeable inserts that hold microtubes, centrifuge tubes, scintillation vials, universals, cryotubes and falcon tubes
  • Also supplied is our range of floor-standing and benchtop bins with anti-slip feet and hinged lids
  • These serve as an ideal solution for short-term storage of radioactive waste or radioisotopes
  • Both the Beta and Gamma storage bins are available in five sizes, while the two largest bin models have wheels for easy transportation
  • Optional heavy duty drawstring bags may also be purchased
Specifications
Specifications
For Maxi Boxes
Insert
160mm
40mm
15 x 15mL