Learn More
Sampling Systems™ Inserts for use with Multi Thief
Enables the sample volume to be quickly and easily changed by changing the removable sample cells. Sampling Systems™ Insert for use with Multi Thief is ideal for taking multiple bulk samples from free flowing powders and granules.
Brand:  Sampling Systems™ 1290W-05
 View more versions of this product
Product Code. 15859241
  • 207.00 € / Each
Estimated Shipment: 04-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • 316 stainless steel construction
  • Variable sample volumes
  • Range of sampler lengths
  • Suitable for free flowing powders and granules
  • Multilevel Powder Sampler with variable Sample Volumes
Specifications
Specifications
Sampler Insert
Stainless Steel (316)
5 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Special Offers
Special Offers
Product Content Correction
Product Title
Sampling Systems™ Inserts for use with Multi Thief > Capacity: 5mL