Learn More
Abeba™ Insoles 3567
ESD dissipative and antistatic insole is flat, smooth and antistatic.
Brand:  Abeba™ 3567/39
 View more versions of this product
Product Code. 15610218
  • 16.40 € / 1 pair
Estimated Shipment: 24-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Microfiber
  • White
  • ESD dissipative according to DIN EN 61340
  • Antistatic
  • Flat, smooth
  • Footbed Reflexor
Specifications
Specifications
White
Microfiber
39
3567