Learn More
Abeba™ Insoles 3570
Massage knobs stimulate circulation. ESD dissipative and antistatic.
Brand:  Abeba™ 3570/43
 View more versions of this product
Product Code. 15620238
  • 16.40 € / 1 pair
Estimated Shipment: 24-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Microfiber
  • Anthracite
  • ESD dissipative according to DIN EN 61340
  • Antistatic
  • Stimulating circulation by massage knobs, comfortable orthopaedic footbed
  • Reflexor massage active-footbed
Specifications
Specifications
Anthracite
Microfiber
43
3570