Learn More
Instrument Calibration(ICAL) Standard, Solution B, Specpure™, Thermo Scientific Chemicals
319.59 € - 913.00 €
Specifications
Chemical Name or Material | Instrument Calibration (ICAL) Standard Solution B |
---|---|
MDL Number | MFCD08064527 |
UN Number | UN1789 |
Grade | Specpure™ |
Physical Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11346269
|
Thermo Scientific Alfa Aesar
044856.AE |
100 mL |
319.59 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11356269
|
Thermo Scientific Alfa Aesar
044856.AP |
500 mL | N/A | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Instrument Calibration (ICAL) Standard Solution B | |
UN1789 | |
Liquid |
MFCD08064527 | |
Specpure™ |
Safety and Handling
GHS H Statement
H314
Causes severe skin burns and eye damage.
P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
H290-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: HYDROCHLORIC ACID
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.