missing translation for 'onlineSavingsMsg'
Learn More

Instrument Calibration Standard 2 For ICP/MS, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Brand:  SPEX Certiprep CL-CAL-2


Product Code. 15628245

  • 539.00 € / 125mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
This item is not returnable. View return policy

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Ag100μg/mL
Al100μg/mL
As100μg/mL
Ba100μg/mL
Be100μg/mL
Ca100μg/mL
Cd100μg/mL
Co100μg/mL
Cr100μg/mL
Cu100μg/mL
Fe100μg/mL
K100μg/mL
Mg100μg/mL
Mn100μg/mL
Mo100μg/mL
Na100μg/mL
Ni100μg/mL
Pb100μg/mL
Sb100μg/mL
Se100μg/mL
Sn100μg/mL
Sr100μg/mL
Ti100μg/mL
Tl100μg/mL
V100μg/mL
Zn100μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Instrument Calibration Standard 2
125 mL
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Each component at 100μg/mL in 5% HNO3/tr. Tartaric Acid/tr. HF:
Aluminum
Antimony
Arsenic
Barium
Beryllium
Cadmium
Calcium
Chromium
Cobalt
Copper
Iron
Lead
Magnesium
Manganese
Molybdenum
Nickel
Potassium
Selenium
Silver
Sodium
Strontium
Thallium
Tin
Titanium
Vanadium
Zinc
Triple-leached LDPE bottle
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Trace ICP/ICP-MS
5% HNO3/tr Tart. Acid/tr HF
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • instrument calibration standard 2

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P362-Take off contaminated clothing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : nitic acid

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product intended for laboratory use only