Learn More
Hanna Instruments™ Chlorine Reagent Sets, For Colorimetric Water Testing
Allows users to achieve fast and accurate colorimetric measurements. Hanna Instruments™ Chlorine Reagent Sets, For Colorimetric Water Testing are high-quality, pre-measured solutions for use with compatible benchtop and/or portable Hanna meters.
134.00 €
Product Code | Brand | For Use With (Application) | No. of Tests | Range | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Application) | No. of Tests | Range | Price | Quantity & Availability | |||
10856340
|
Hanna Instruments™
HI9373403 |
Free and total chlorine high range determination | 300 | 0.00 to 10.00mg/L free and total chlorine |
134.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
These high quality reagents are designed to follow accepted test methods for colorimetric determinations of total and/or free chlorine. By simply adding the reagents to the sample as directed, a reaction will take place and a compatible meter can be used to determine the concentration from the color that is produced.
- Pre-made, pre-measured water testing reagents for ease of use
- Prepared with high purity chemicals manufactured in Hanna's state-of-the-art facility
- Clearly marked with expiration date and lot number on each packet and bottle for traceability
- Reagent sets available for: Free and total chlorine (high range), Free chlorine (ultra low range), Total chlorine (ultra high range, ultra low range, undetermined range)
Specifications
1 Set | |
Water Testing Kit |