Learn More
Tocris Bioscience™ Insulin (human) recombinant, expressed in yeast
Endogenous peptide agonist
Brand:  Tocris Bioscience™ 3435/10
100.04 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18065782
  • 128.00 € / 10mg
Explore more special offers
Description
Description
Insulin (human, recombinant, expressed in yeast) is an endogenous insulin receptor agonist (Ki = 4.85 nM). Decreases plasma glucose levels, proteolysis, lipolysis and gluconeogenesis and increases glycogen and fatty acid synthesis in vivo.