missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin beta-7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11328-1-AP-20UL
This item is not returnable.
View return policy
Description
Integrin beta 7 associates with integrin alpha 4 (CD49d) to form the alpha 4 beta 7 integrin LPAM-1, expressed on intraepithelial lymphocytes. It also associates with alpha E (CD103) to form the alpha E beta 7 integrin HML-1, expressed on T cells adjacent to mucosal epithelium and intraepithelial lymphocytes. Ligands for LPAM-1 include VCAM-1 (CD106), MAdCAM-1 and fibronectin, while the ligand of HML-1 is E-cadherin (CD324). Integrin beta 7 plays an important role in the adhesion of leukocytes to endothelial cells promoting the transmigration of leukocytes to extravascular spaces during the inflammatory response.Specifications
Integrin beta-7 | |
Polyclonal | |
Unconjugated | |
ITGB7 | |
Integrin beta 7, integrin, beta 7, ITGB7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
16421, 25713, 3695 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P26010, P26011 | |
ITGB7 | |
Integrin beta-7 Fusion Protein Ag1871 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |