missing translation for 'onlineSavingsMsg'
Learn More

Interference Check Standard 18 without Mercury For Environmental Analysis, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

382.00 € - 662.00 €

Specifications

Chemical Name or Material Interference Check Std 18 without Mercury
Packaging Triple-leached LDPE bottle
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Grade Environmental
View More Specs

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15296096
View Documents
SPEX Certiprep
INTER18-100N
125 mL Triple-leached LDPE bottle
382.00 €
125mL
Please to purchase this item. Need a web account? Register with us today!
15206106
View Documents
SPEX Certiprep
INTER18-500N
500 mL Triple-leached LDPE bottle
662.00 €
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
K20,000μg/mL
As1000μg/mL
Pb1000μg/mL
Tl1000μg/mL
Se500μg/mL
Ag300μg/mL
Ba300μg/mL
Cd300μg/mL
Co300μg/mL
Cr300μg/mL
Cu300μg/mL
Ni300μg/mL
V300μg/mL
Zn300μg/mL
Mn200μg/mL
Be100μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Interference Check Std 18 without Mercury
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Environmental
5% HNO3
Triple-leached LDPE bottle
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Various
Videos
Safety and Handling

Safety and Handling

Causes severe skin burns and eye damage

missing translation for 'dotInformation' : Hazardous

Recommended Storage : Ambient

Product Identifier
  • Interference Check Standard, 18

Signal Word
  • Danger

Hazard Category
  • Carcinogenicity Category 1A
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 1A
  • Skin corrosion/irritation Category 1 A

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H350-May cause cancer.
  • H360-May damage fertility or the unborn child.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P363-Wash contaminated clothing before reuse.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : nitric acid, potassium, lead, thallium,arsenic

SDS
Documents

Documents

Product Certifications

Product intended for laboratory use only