missing translation for 'onlineSavingsMsg'
Learn More

Internal Standard, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

107.00 € - 113.00 €

Specifications

Chemical Name or Material Internal Standard
Name Note EPA Method 625, SW-846 Method 8270C, CLP Methods
Packaging 2mL amber prescored glass ampule
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
12769705
View Documents Promotion Details
SPEX Certiprep
CLPS-I
1 mL 2mL amber prescored glass ampule
107.00 €
1mL
Please to purchase this item. Need a web account? Register with us today!
12779705
View Documents Promotion Details
SPEX Certiprep
CLPS-I2
2 mL 2mL amber prescored glass ampule This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
1,4-Dichlorobenzene-d43855-82-14000μg/mL
Acenaphthene-d1015067-26-24000μg/mL
Chrysene-d121719-03-54000μg/mL
Naphthalene-d81146-65-24000μg/mL
Perylene-d121520-96-34000μg/mL
Phenanthrene-d101517-22-24000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Internal Standard
2mL amber prescored glass ampule
GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
For US EPA Methods 625, 8270C, CLP
EPA Method 625, SW-846 Method 8270C, CLP Methods
Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
4000μg/mL
MeCl
Videos
Safety and Handling

Safety and Handling

missing translation for 'dotInformation' : Non-Hazardous

Recommended Storage : Ambient

SDS
Documents

Documents

Product Certifications

Product intended for laboratory use only