missing translation for 'onlineSavingsMsg'
Learn More

Labnet™ Mini LabRoller Rotator with Rotisserie

Simple, yet versatile, end-over-end tube rotator. Labnet™ Mini LabRoller Rotator with Rotisserie is designed for mixing samples in both horizontal and vertical planes.

Brand:  Labnet™ H5500-230V-EU

Product Code. 11895953

  • 446.00 € / Each
Estimated Shipment: 17-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Compact design
  • Wide selection of motions
  • Noiseless operation
  • Cold room compatible
  • Dual format rotisseries
  • Includes rotisserie H5500-02
  • Other rotisseries are available (sold separately): H5500-15, H5500-50
  • Holds various size tubes:
    • 36 x 1.5 / 2.0mL
    • 10 x 15mL and 12 x 5mL
    • 6 x 50mL
    • 2 x 96 well plates
  • Can be installed inside the I5110A-230V-UK Labnet incubator
Specifications

Specifications

230 V, 50/60 Hz
50/60 Hz
Rotator with one Rotisserie
230 V
21.3 x 10.2 x 12.6 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers