missing translation for 'onlineSavingsMsg'
Learn More

3M™ E-A-R™ E-A-Rsoft Yellow Earplugs

Product Code. p-8010115
Click to view available options
Corded:
Corded
Uncorded
Unit Size:
1 pair
200 pairs
250 pairs
This item is not returnable. View return policy

Product Code. 12364749

Brand: 3M™ 7100111802

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Offers high attenuation and are made from a slow expanding, PU foam material. 3M E-A-R Polyurethane Earplug – 36dB.

  • Made from a slow expanding, PU foam material
  • Provides evenly distributed pressure, giving flexibility, and a good seal with optimum comfort.
  • Bright, neon color makes it appealing to wearers
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow Neon
Certifications/Compliance CE Approved
For Use With (Application) For general industrial use
Pairs per Package 250 Pairs
Noise Reduction Rating (NRR) 36 dB
Corded Uncorded
Product Type Ear Plugs
Material Polyurethane
Style Roll-down
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.