missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 100 bp DNA Ladder
GREENER_CHOICE

Invitrogen 100 bp DNA Ladder is designed for sizing and approximate quantification of double-stranded DNA in the range of 100 bp to 2,000 bp.

Brand:  Invitrogen™ 15628050

247.34 EUR valid until 2024-09-28
Use promo code "21626" to get your promotional price.


 View more versions of this product

Product Code. 11580916

  • 371.00 € / 250µg
In Stock
to see stock.
Explore more special offers
Add to basket

Alert:

To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price

This item is not returnable. View return policy

Description

Description

Invitrogen 100 bp DNA Ladder is designed for sizing and approximate quantification of double-stranded DNA in the range of 100 bp to 2,000 bp. 100 bp DNA Ladder consists of 13 individual chromatography-purified DNA fragments and has reference bands at 2000, 1500, and 600 bp for easy orientation.

100 bp DNA Ladder is ideal for separation on 1–2% agarose gels.

Highlights of 100 bp DNA Ladder:
Sharp, clear bands—chromatography purified fragments for consistent and reliable results
Convenient—provided with 10X BlueJuice Gel Loading Buffer for tracking of sample DNA migration
Precise—an exact amount of DNA in each band

Product use
The double-stranded DNA ladder can be visualized on 1–2% agarose gels after ethidium bromide or SYBR Safe staining. The ladder is designed with a uniform intensity of DNA bands for a clear view of each band. An exact amount of DNA in each band allows approximate quantification of DNA samples.

This ladder can be radiolabeled with T4 polynucleotide kinase or T4 DNA polymerase.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

• 500 µL 100 bp DNA Ladder
• 1 mL 10X BlueJuice Gel Loading Buffer

Store at -20°C.

Agarose
No
100 to 2000 bp
Agarose gel
500 Applications
Approved for shipment at Room Temperature or on Dry Ice
0.5 μg/μL
DNA Ladder
1 mL
500 μL
Sustainable packaging
500 μL DNA Ladder, 1 mL 10X Loading Buffer
Individual chromatography-purified DNA fragments
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.