missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 2-YT Broth

2-YT Broth is a nutrient-rich microbial growth medium used for cultivation of E. coli and growth of hosts for replication of M13 vectors.

Brand:  Invitrogen™ 22712020

120.08 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.


Product Code. 11548916

  • 160.00 € / 500g

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

2-YT Broth is a nutrient-rich microbial growth medium used for cultivation of E. coli and growth of hosts for replication of M13 vectors. The 2-YT formulation is similar to Lennox LB medium, but contains double the concentration of yeast extract and a 60% higher concentration of peptone. 2-YT medium allows cells to reach higher density and increase recombinant protein production compared to LB media formulations.

2-YT Broth formulation
Formulation per liter: 16 g SELECT Peptone 140 (pancreatic digest of casein), 10 g SELECT Yeast Extract, and 5 g NaCl.
Note: To prepare 1 L of 2-YT medium, weigh 31 g of powder and add distilled water to 1 L. Adjust the pH value to 7.0 with 5 N NaOH and sterilize by autoclaving for 20 minutes at 121°C.

Primary applications
• Growth of hosts for replication of M13 vectors
• Recombinant protein expression
• Achieve higher plasmid DNA yield

Find the media format that fits you needs
We offer a variety of ready-to-use media formulations including LB Broth (Cat. No. 10855001), Terrific Broth (Cat. No. A1374301) and S.O.C Recovery Medium (Cat. No. 15544034).

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Escherichia coli
Bottle
Autoclave
Coliforms
Room Temperature
• 2-YT Broth Powder (500 g)

Store at room temperature.

2-YT Broth (Yeast Extract Tryptone)
Dehydrated Culture Media
500 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2110CU Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number22712020
Lot Number2210BF Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number22712020
Lot Number2302CB Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number22712020
Lot Number2301BK Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number22712020
Lot Number2308AP Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number22712020
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ 2-YT Broth > 500g

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.