missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 95401.H4POOL (Yeast Deletion Pools)

Allow simultaneous analysis of large numbers of deletion strains through selective growth conditions

Brand:  Invitrogen™ 95401.H4POOL

Product Code. 10722734

  • 7260.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: Supplied in five tubes, each containing 200μL of the pooled deletions.
This item is not returnable. View return policy

Description

Description

  • Identification and quantitation of deletions surviving selection can be determined by hybridizing deletions to microarray containing all the barcode oligos
  • Barcode oligos are unique to each deletion, allowing rapid, high-throughput identification of deletions
  • Each Yeast Deletion Pool contains all deletions within strain, equally represented within pool

Gene Expression Analysis & Genotyping, Genotyping & Genomic Profiling, Yeast Deletion Mapping

Order Info

Shipping Condition: Dry ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Supplied in five tubes, each containing 200μL of the pooled deletions
-68°C to -85°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.