missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 5-(3-Aminoallyl)-dUTP (50 mM)

When incorporated into DNA, Ambion 5-(3-aminoallyl)-dUTP provides a reactive group for addition of other chemical groups.

Brand:  Invitrogen™ AM8439

181.20 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.


Product Code. 10526735

  • 302.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

When incorporated into DNA, Ambion 5-(3-aminoallyl)-dUTP provides a reactive group for addition of other chemical groups. The modified dUTP is readily incorporated by reverse transcriptase. The aminoallyl modification enables downstream reaction with amine-reactive compounds such as N-hydroxysuccinimide (NHS) esters; thus aminoallyl-modified cDNA can be labeled with any amine-reactive group moiety.

Use of this modified nucleotide
Use 5-(3-aminoallyl)-dUTP as recommended in your protocol for reverse transcription. Substitution of 30–80% of the dTTP with 5-(3-aminoallyl)-dUTP may be useful, but the exact proportion will depend on experimental goals.

Order Info

Shipping Conditions: Dry Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

5-(3-Aminoallyl)-dUTP, 50 mM (50 μL)

Store at –70°C.
Aminoallyl
Nucleotides
50 μL
Nucleic Acid Labeling
Liquid
Ambion™
HPLC
Dry Ice
50 mM
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.