missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 5-(3-Aminoallyl)-UTP (50 mM)

Ambion modified nucleotides confer unique characteristics to the RNA molecules into which they are incorporated.

Brand:  Invitrogen™ AM8437

177.60 EUR valid until 2024-12-20
Use promo code "21651" to get your promotional price.


Product Code. 10733447

  • 296.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Ambion modified nucleotides confer unique characteristics to the RNA molecules into which they are incorporated. Incorporation of an aminoallyl-modified NTP, for example, allows for the subsequent coupling of the reactive amino group to another molecule such as biotin or a Cy dye. Unlike NTPs directly coupled to a dye, aminoallyl-modified NTPs are incorporated almost as efficiently as unmodified NTPs and are much less expensive than dye-coupled NTPs.

Ambion 5-(3-Aminoallyl)-UTP (50 mM) is supplied in a single tube at 50 mM.

Order Info

Shipping Conditions: Dry Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Nucleotides
Dry Ice
Aminoallyl
Ambion™
Liquid
5-(3-Aminoallyl)-UTP, 50 mM (50 μL)

Store at –70°C.
50 mM
50 μL
Nucleic Acid Labeling
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.