missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ siPORT™ NeoFX™ Transfection Agent with manual

Lipid-based formulation can be used to efficiently transfect adherent cells with siRNA as they are subcultured—without increased cytotoxicity

Brand:  Invitrogen™ AM4511M

 View more versions of this product

Product Code. 10005124

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers

Includes: One tube containing 0.4mL
This item is not returnable. View return policy

Description

Description

  • Versatile-works with a broad range of cell lines
  • Reproducible-produces consistent results, lot-to-lot, plate-to-plate, and well-to-well
  • Efficient-performs with high siRNA transfection efficiency, allowing use of low siRNA concentrations to minimize nonspecific effects
  • Fast-reverse-transfects cells as they are plated, saving a day
  • Reverse-Transfect Cells as They are Plated, Saving a Day
  • Optimized for Lower siRNA Concentrations and High Reproducibility
  • Compatible with a Broad Range of Cell Lines

Cell Culture, RNAi Transfection, Synthetic siRNA Transfection, Transfection

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store at 4°C.
6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks
Lipid-based Transfection
High-throughput Compatible
Transfection Reagent
Yes
Stem Cells, Primary Cells, Mammalian
Synthetic siRNA
Transfection
Ambion™, siPORT™
1 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.