missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Bac-to-Bac™ Baculovirus Expression System
Enables efficent production of recombinant baculovirus for expression testing in insect cells
Brand: Invitrogen™ 10359016
1363.40 EUR valid until 2025-03-28
Use promo code "24107" to get your promotional price.
Includes: Each Bac-to-Bac Baculovirus Expression System includes 10μg of pFastBac1 vector, 0.5mL MAX Efficiency DH10Bac Chemically Competent Cells, pFastBac 1-Gus control vector, and 1mL of Cellfectin Reagent.
Description
- Relies on generation of recombinant baculovirus by site-specific transposition in E. colirather than homologous recombination in insect cells
- Time-saving expression bacmid: expression cassette of pFastBacvector recombines with parent bacmid in DH10BacE. coli
- Competent Cells to form expression bacmid
- Bacmid is then transfected into insect cells for production of recombinant baculovirus particles
- Easy colony screening
- Parent bacmid in DH10BacE. colicontains segment of lacZα gene
- lacZα gene is disrupted upon transposition of expression cassette into bacmid allowing for blue/white selection of recombinants
- Flexibility
- Comes with pFastBac1 vector that offers strong polyhedrin promoter for protein expression and large multiple cloning site for simplified cloning
- Two other vectors are offered separately
- pFastBacHT vector for production of histidine-tagged recombinant protein and pFastBacDual vector for expression of two proteins simultaneously
- Rapid purification of recombinant proteins
- Optional pFastBacHT vector produces histidine-tagged recombinant protein for rapid protein purification with nickel-chelating resin
Baculovirus Production and Titering, Insect Expression, Protein Expression, Proteins, Expression, Isolation and Analysis
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Polyhedrin | |
Each Bac-to-Bac Baculovirus Expression System includes: • pFastBac1 vector (10 μg), store at -20°C • pFastBac 1-Gus control vector (4 ng), store at -20°C • MAX Efficiency DH10Bac Chemically Competent E. coli (1 kit with 5 x 0.1 mL), store at -80°C • ExpiFectamine Sf Transfection Reagent (1 mL), store at 4°C, do not freeze |
|
Restriction Enzyme/MCS | |
pFastBac |
Baculovirus Expression System | |
Untagged | |
1 kit | |
Bac-to-Bac™ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction