missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BaculoDirect™ C-Term Expression Kit
Have attR sites for recombination with any attL-flanked fragment, regardless of whether it is entry clone or Ultimate™ RF Clone
Brand: Invitrogen™ 12562013
2057.00 EUR valid until 2025-03-28
Use promo code "24107" to get your promotional price.
Includes: All destination vectors are provided lyophilized and supercoiled.
Requires : Gateway™ entry clone, Gateway™ LR Clonase enzyme mix, and reaction buffer.
This item is not returnable. View return policy
Description
Baculovirus Production and Titering, Cloning, Gateway Cloning, Insect Expression, Protein Expression, Proteins, Expression, Isolation and Analysis

Specifications
Polyhedrin | |
The BaculoDirect expression kits include five 0.3 µg vials of BaculoDirect Linear DNA, 125 µL of Cellfectin transfection reagent, frozen Sf9 cells, Gateway LR Clonase enzyme mix, GIBCO Unsupplemented Grace's Insect Medium, and ganciclovir. Store the BaculoDirect Linear DNA, Cellfectin reagent, and Grace's Medium at +4°C. Store LR Clonase mix at -80°C. Store Sf9 cells in liquid nitrogen. All components are guaranteed stable for 6 months if stored properly. |
|
Gateway | |
BaculoDirect Vectors |
Expression Kit | |
His Tag (6x), V5 Epitope Tag | |
5 transfections | |
BaculoDirect™, Gateway™ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Invitrogen™ BaculoDirect™ C-Term Expression Kit > 5 transfections
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction