Learn More
Invitrogen™ BLOCK-iT™ Lentiviral RNAi Gateway™ Vector Kit
The pLenti6/BLOCK-iT-DEST expression vector provided in the BLOCK-iT Lentiviral RNAi Expression System can be used to efficiently introduce and stably express short hairpin RNA (shRNA) in vivo from a lentiviral vector
Brand: Invitrogen™ K494300
Includes: The BLOCK-iT™ Lentiviral RNAi Gateway™ Vector Kit contains the pLenti6/BLOCK-iT™-DEST vector, pLenti6-GW/U6-laminshRNA control vector, and One Shot™ Stbl3 Competent Cells. Vectors are supplied supercoiled and lyophilized. The BLOCK-iT™ Lentiviral RNAi Expression System contains the BLOCK-iT™ Lentiviral RNAi Gateway™ Vector Kit, BLOCK-iT™ U6 Entry Vector Kit, ViraPower™ Bsd Support Kit, 293FT Cell Line, and LR Clonase™ enzyme mix.
Description
A novel cloning process places an approx. 50-bp DNA oligonucleotide immediately following a U6 pol III promoter into the BLOCK-iT U6 entry vector. The oligonucleotide is designed to express RNA that forms a stemLoop structure containing the sense and antisense regions of your target gene of interest. This shRNA is then recombined into the pLenti6/BLOCK-iT-DEST vector. After viral production and transduction, the shRNA driven by the U6 promoter becomes stably integrated as an RNAi cassette. The shRNA generated avoids the hosts defense mechanism and will be effective at producing the RNAi gene knockdown response. Using the BLOCK-iT Lentiviral RNAi Expression System, long-term analysis of gene blocking in both dividing and non-dividing mammalian cell types and animal models can be achieved.
The BLOCK-iT RNAi U6 Entry Vector Kit allows streamlined cloning of shRNA target sequences for testing in transient experiments. Selected RNAi expression cassettes are quickly and efficiently recombined from the BLOCK-iT RNAi U6 entry vector into the pLenti6/BLOCK-iT-DEST vector via a standard Gateway LR recombination reaction.
- attR sites for efficient recombination with the attL-flanked U6 Gateway™ entry vector containing the RNAi cassette
- All of the required components for efficient lentiviral packaging and delivery of the shRNA of interest
- Blasticidin selection marker for fast, efficient selection of stable cell lines expressing the shRNA
Cloning, Gateway Cloning, Lentiviral shRNA and miRNA, RNAi, RNAi, Epigenetics and Non-Coding RNA Research, Virus-Based RNAi
Specifications
RNAi Expression Vector Kit | |
Gateway™ | |
Lentiviral | |
Blasticidin | |
U6 | |
BLOCK-iT™, Gateway™ |
The BLOCK-iT™ Lentiviral RNAi Gateway™ Vector Kit contains the pLenti6/BLOCK-iT™-DEST vector, pLenti6-GW/U6-laminshRNA control vector, and One Shot¤ Stbl3 Competent Cells. Vectors are supplied supercoiled and lyophilized. Store at -20°C once resuspended. Store Competent Cells at -80°C. The BLOCK-iT™ Lentiviral RNAi Expression System contains the BLOCK-iT™ Lentiviral RNAi Gateway™ Vector Kit, BLOCK-iT™ U6 Entry Vector Kit, ViraPower™ Bsd Support Kit, 293FT Cell Line, and LR Clonase™ enzyme mix. Store components as indicated in the respective kits. Store the LR Clonase™ mix at -80°C. Guaranteed stable for 6 months when properly stored. | |
Constitutive | |
shRNA | |
pLenti | |
Kit | |
20 Reactions |
For Research Use Only. Not for use in diagnostic procedures.