missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BlueJuice™ Gel Loading Buffer (10X)
GREENER_CHOICE

Designed for easy loading and tracking of DNA samples in agarose or native polyacrylamide gels

Brand:  Invitrogen™ 10816015

Product Code. 10666143

  • 43.68 € / Pack of 3
In Stock
to see stock.
Explore more special offers
Add to basket

Includes: 3 vials BlueJuice™ Gel Loading Buffer (10X), 1mL each
This item is not returnable. View return policy

Description

Description

  • Designed for easy loading and tracking of DNA samples in agarose or native polyacrylamide gels
  • Recommended concentration for use with DNA samples run on agarose gels is 2X (one part buffer plus four parts sample)R
  • recommended concentration is 1X for acrylamide gels

Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

• 3 x 1 mL 10X BlueJuice Gel Loading Buffer

Store at room temperature or at 4°C for up to 6 months. For longer periods, store at -20°C.

10X
Bromophenol Blue
1 mL
BlueJuice™
Room Temperature
Sample Loading Buffers
Agarose
Gel Loading Buffer
Agarose Gels
3 x 1 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.