missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Bluo-Gal

Bluo-Gal (halogenated indolyl-β-galactoside) is an alternative histochemical substrate for β-galactosidase that produces a darker blue color than X-gal, the more frequently used substrate.

Brand:  Invitrogen™ 15519028

493.24 EUR valid until 2024-12-20
Use promo code "21651" to get your promotional price.


Product Code. 10133862

  • 649.00 € / 1g
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Bluo-Gal (halogenated indolyl-β-galactoside) is an alternative histochemical substrate for β-galactosidase that produces a darker blue color than X-gal, the more frequently used substrate. As with X-gal, Bluo-Gal produces blue (lac+) bacterial colonies and blue (lac+) viral plaques. Bacteria containing active β-galactosidase produce blue colonies when grown on plates containing Bluo-Gal.

Bluo-Gal can also be used as a substrate with the streptavidin-β-galactosidase conjugate in both immunoblot and immunocytochemistry assays. In the presence of β-galactosidase, Bluo-Gal yields a blue precipitate, which is insoluble in alcohols and xylenes. Bluo-Gal is soluble in dimethylformamide.

Primary applications
• Blue-white screening (deeper blue as compared to X-gal)—detect functional β-galactosidase (reporter gene expression)
• Histochemical detection of β-galactosidase activity in fixed tissues
• Bluo-Gal overlay assay

Find the media format that fits your needs
• Ready-to-use media formulations are available, such as LB Broth (Cat. No. 10855001), Terrific Broth (Cat. No. A1374301), or S.O.C recovery medium (Cat. No. 15544034).
• IPTG (Cat. No.15529019) is available for blue-white screening.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

• Bluo-Gal, substrate for β-galactosidase (1 g)

Store at –5°C to –30°C.

Colorimetric
Blue/White Colony Screening, Immunoblotting, Immunocytochemical Assays
1 g
Bluo-Gal, Cell-Permeant
Bottle
Halogenated Indolyl-β-galactoside Substrate
Approved for shipment at Room Temperature or on Wet Ice
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.