Learn More
Invitrogen™ Champion™ pET SUMO Expression System
Yield high levels of protein expression in E. coli
Brand:  Invitrogen™ K30001
901.68 EUR valid until 2024-12-20
Use promo code "21651" to get your promotional price.
Product Code. 10144802
  • 1156.00 € / Each
Explore more special offers
Includes: The Champion pET SUMO TA Cloning box contains linearized Champion pET SUMO vector, sterile water, dNTPs, 10X PCR Buffer, control template and primers, T4 DNA ligase, 10X ligation buffer, primers for sequencing or PCR screening, and an expression control. The SUMO Protease box contains SUMO Protease and buffers. The TOP10 box contains 21 x 50μL aliquots of chemically competent E. coli, S.O.C. medium, and a control plasmid. The One Shot BL21(DE3) box contains 21 x 50μL aliquots of chemically competent E. coli, S.O.C. medium, and a control plasmid.
Description
Description
  • Uses small ubiquitin-related modifier (SUMO) fusion, belonging to growing family of ubiquitin-related proteins, to enhance solubility of expressed fusion proteins
  • Contrast to ubiquitin, SUMO is involved in stabilization and localization of proteins in vivo
  • After expression, 11 kd SUMO moiety can be cleaved by highly specific and active SUMO (ULP-1) protease at carboxyl terminal, producing native protein
  • Enhanced solubility with N-terminal SUMO fusion
  • Highly efficient cleavage produces native protein of interest with SUMO (ULP-1) protease
  • Highly specific cleavage eliminates chance of protein of interest being internally digested, regardless of its amino acid sequence
  • Significantly increased stability with SUMO fusion-can be used for small peptide productionT7lac promoter for high-level protein expression
  • N-terminal 6xHis tag for protein detection and purification
Bacterial Expression, Protein Expression, Proteins, Expression, Isolation and Analysis
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Inducible
T7, lacO
None
Kanamycin (KanR)
SUMO
Cell-Based Expression
TOPO
pET
IPTG
Expression System
The Champion™ pET SUMO Expression System is provided as a complete system. The Champion™ pET SUMO TA Cloning™ box contains linearized Champion™ pET SUMO vector, sterile water, dNTPs, 10X PCR Buffer, control template and primers, T4 DNA ligase, 10X ligation buffer, primers for sequencing or PCR screening, and an expression control. Store at -20°C. The SUMO Protease box contains SUMO Protease and buffers. Store at -80°C. The One Shot™ TOP10 box contains twenty-one 50 μl aliquots of chemically competent E. coli, S.O.C. medium, and a control plasmid. Store at -80°C. The One Shot¤ BL21(DE3) box contains twenty-one 50 μl aliquots of chemically competent E. coli, S.O.C. medium, and a control plasmid. Store at -80°C.
BL21(DE3)
E. coli
His Tag (6x), SUMO Tag
20 reactions
Champion™, TA Cloning™