missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Champion™ pET101 Directional TOPO™ Expression Kit with BL21 Star™ (DE3) One Shot™ Chemically Competent E. coli
Yields protein of interest at levels greater than 50 percent of total cellular protein in E. coli
Brand: Invitrogen™ K10101
916.30 EUR valid until 2024-09-27
Use promo code "21650" to get your promotional price.
Includes: 200ng of linearized, topoisomerase I-activated Champion pET vector; sterile water; dNTPs; 10X PCR Buffer; salt solution; control template and primers; primers for sequencing or PCR screening; and an expression control. The TOP10 box contains twenty-one 50μL aliquots of chemically competent E. coli, S.O.C. medium, and a control plasmid.
Description
- Based on T7 expression vectors originally developed by Studier and colleagues, high-level expression is achieved because T7 RNA polymerase is more processive than native E. coliRNA polymerase and is dedicated to transcription of gene of interest
- Protein production is further enhanced in system by expression strain BL21 StarE. coli, which significantly improves stability of mRNA transcripts and increases protein expression up to ten-fold
- Simplified, efficient Directional TOPO cloning allows to quickly enter ChampionpET Expression vector
- Features linearized, topoisomerase I-activated ChampionpET expression vectors for five-minute directional cloning
- Directional TOPO Cloning technology facilitates gene expression becausa proofreading enzyme is used for PCR, resulting in fewer errors in cloned genes
- Greater than 90% of clones are in correct orientation for gene expression, reducing time spent on colony screening
Bacterial Expression, Protein Expression, Proteins, Expression, Isolation and Analysis
Specifications
Inducible | |
T7, lacO | |
None | |
Ampicillin (AmpR) | |
E. coli | |
His Tag (6x), V5 Epitope Tag | |
20 reactions | |
One Shot™ |
IPTG | |
TOPO Expression Kit | |
Each Champion™ pET Directional TOPO™ Expression Kit is provided as a complete expression system. The Directional TOPO™ Expression box contains 200 ng of linearized, topoisomerase I-activated Champion™ pET vector; sterile water; dNTPs; 10X PCR Buffer; salt solution; control template and primers; primers for sequencing or PCR screening; and an expression control. Store at -20°C. The One Shot™ TOP10 box contains twenty-one 50-μl aliquots of chemically competent E. coli, S.O.C. medium, and a control plasmid. Store at -80°C. The One Shot™ BL21 Star™(DE3) box contains twenty-one 50-μl aliquots of chemically competent E. coli, S.O.C. medium, and a control plasmid. Store at -80°C. Kits with Lumio™ Technology include 20 μl of the Lumio™ Detection Reagent. Store at -20°C. Guaranteed stable for 6 months when properly stored. | |
BL21 Star™(DE3) | |
Cell-Based Expression | |
Directional TOPO | |
pET |
For Research Use Only. Not for use in diagnostic procedures.