missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Gateway™ BP Clonase™ Enzyme Mix

Brand:  Invitrogen™ 11789021

2313.30 EUR valid until 2024-12-20
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 10003152

  • 3505.00 € / 100 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: Proteinase K solution (2μg/μL), 30% PEG 8000/30mM MgCl2 Solution and a positive control vector.
This item is not returnable. View return policy

Description

Description

  • Proteins catalyze in vitro recombination of PCR products or DNA segments from clones (containing attB sites) and Donor vector (containing attP sites) to generate Entry clones
  • BP Clonase II enzyme format is single mix of both enzyme and buffer to ensure enzyme stability and enable convenient 10μL reaction set up with fewer pipetting steps, which both maximize recombination efficiency and minimize pippetting
  • Original format of BP Clonase enzyme is stand alone enzyme blend and reaction buffer provided in separate tubes

Cloning, Gateway Cloning, Gateway Recombination

Order Info

Shipping Conditions: Dry ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

All Gateway™ BP Clonase™ enzyme kits include proteinase K solution (2 μg/μl), 30% PEG 8000/30 mM MgCl2 Solution and a positive control vector. Store BP Clonase™ enzyme at -80°C; BP Clonase™ II Enzyme mixtures at -20°C. Guaranteed stable for 6 months when properly stored.
BP Clonase
100 Reactions
Clonase™, Gateway™
Dry Ice
Reaction Buffer
BP Clonase Enzyme Mix
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.